Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810218P

Avanti

C6-NBD Sphingomyelin

Avanti Research - A Croda Brand 810218P, powder

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-sphingosine-1-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$697.31

$697.31


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$697.31

About This Item

Empirical Formula (Hill Notation):
C35H61N6O9P
CAS Number:
Molecular Weight:
740.87
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$697.31


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810218P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810218P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](NC(=O)CCCCCNc1c2n[o]nc2c(cc1)[N+](=O)[O-])[C@H](O)\C=C\CCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C35H61N6O9P/c1-5-6-7-8-9-10-11-12-13-14-15-16-18-21-32(42)30(28-49-51(46,47)48-27-26-41(2,3)4)37-33(43)22-19-17-20-25-36-29-23-24-31(40(44)45)35-34(29)38-50-39-35/h18,21,23-24,30,32,42H,5-17,19-20,22,25-28H2,1-4H3,(H2-,36,37,39,43,46,47)/b21-18+/t30-,32+/m0/s1

InChI key

YDBDYTGWMRUUSN-PERJAUJZSA-N

General description

Sphingomyelin is a membrane phospholipid with a central sphingosine and an amide linked fatty acid. Phosphocholine forms the polar head group.[1] It is one of the major phospholipid present in milk fat globule membrane.[2]

Application

C6-NBD sphingomyelin is suitable for endocytosis assay.[3]

Biochem/physiol Actions

Sphingomyelin controls cell growth, differentiation, apoptosis and signal transduction along the membrane. It is known to prevent the proliferation of colon tumour. Sphingomyelin also inhibits cholesterol absorption.[2] Abnormal accumulation of sphingomyelin causes Niemann Pick disease.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810218P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioactive Components of Milk, 22-22 (2007)
Apoptosis: Biology and Mechanisms, 66-66 (2013)
mTOR complex 1 activity is required to maintain the canonical endocytic recycling pathway against lysosomal delivery
Dauner K, et al.
The Journal of biological chemistry, 292(14), 5737-5747 (2017)
Mark L Schultz et al.
BMC medicine, 17(1), 200-200 (2019-11-13)
Niemann-Pick disease type C is a fatal and progressive neurodegenerative disorder characterized by the accumulation of unesterified cholesterol in late endosomes and lysosomes. We sought to develop new therapeutics for this disorder by harnessing the body's endogenous cholesterol scavenging particle
Avery L McIntosh et al.
Lipids, 45(6), 465-477 (2010-05-18)
During the last several years, intracellular lipid droplets have become the focus of intense study. No longer an inert bystander, the lipid droplet is now known as a dynamic organelle contributing lipids to many cellular events. However, while the dynamics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service