Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810213P

Avanti

C12-NBD Sphinganine

Avanti Research - A Croda Brand 810213P, powder

Synonym(s):

N-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-sphinganine

Sign Into View Organizational & Contract Pricing

Select a Size

810213P-500UG
$220.00
810213P-1MG
$386.00

$220.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
810213P-500UG
$220.00
810213P-1MG
$386.00

About This Item

Empirical Formula (Hill Notation):
C36H63N5O6
CAS Number:
Molecular Weight:
661.92
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$220.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (810213P-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810213P

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

C12-Sphinganine is an active derivative of sphinganine (dihydrosphingosine), with C12 being the number of carbon atoms in the acyl residue.[1] C12-NBD Sphinganine is an omega fatty acid labeled fluorescent sphingolipid.

Packaging

5 mL Amber Glass Screw Cap Vial (810213P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Thevissen et al.
Bioorganic & medicinal chemistry letters, 18(13), 3728-3730 (2008-06-06)
The minimal fungicidal concentration (MFC) of dihydrosphingosine (DHS), phytosphingosine (PHS), and five short-chain DHS derivatives was determined for Candida albicans and Candida glabrata. In this respect, a C15- and a C17-homologue of DHS showed a 2- to 10-fold decreased MFC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service