Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810194C

Avanti

18:1-06:0 NBD PS

Avanti Research - A Croda Brand 810194C

Synonym(s):

1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphoserine (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$710.70

$710.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$710.70

About This Item

Empirical Formula (Hill Notation):
C36H61N6O13P
CAS Number:
Molecular Weight:
816.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$710.70


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810194C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810194C

concentration

1 mg/mL (810194C-1mg)

shipped in

dry ice

storage temp.

−20°C

Application

18:1-06:0 NBD PS has been used as a fluorescent analog to observe the lipid transfer from mitochondria to newly autophagosome[1] and to determine the lipid scrambling process.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810194C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cryo-EM structures and functional characterization of the murine lipid scramblase TMEM16F
Alvadia C, et al.
eLife, 8 (2019)
Mitochondria Supply Membranes for Autophagosome Biogenesis during Starvation
Hailey DW, et al.
Cell, 141(4), 656-667 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service