Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

780200C

Avanti

16:0 PE MCC

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (sodium salt), chloroform

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$1,020.00

$1,020.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$1,020.00

About This Item

Empirical Formula (Hill Notation):
C49H86N2O11PNa
CAS Number:
Molecular Weight:
933.18
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,020.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (780200C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 780200C

concentration

10 mg/mL (780200C-25mg)

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

General description

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (16:0 PE MCC) is a lipid that comprises phosphoethanolamine linked to two palmitic acid by its phosphate group and to a maleimide moiety through its amino group. The maleimide group is a thiol acceptor and 16:0 PE MCC is a maleimide-based lipidating reagent.[1]

Application

16:0 PE MCC (1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-4-(p-maleimidomethyl)cyclohexane-carboxamide) may be used:
  • in the preparation of liposomes for conjugation with human immune deficiency virus (HIV-1) envelope glycoprotein trimers[2]
  • in the preparation of oligonucleotide-modified vesicles[3]
  • in the functionalization of the flat silicon wafers[4]
  • in lipid vesicle preparation[5]

Biochem/physiol Actions

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide](1,2-Bis(diphenylphosphino)ethane (DPPE)-MCC) in functionalized liposomes aids in covalent attachment via the maleimide group. It acts as a vehicle in peptide transport studies.[6]

Packaging

5 mL Clear Glass Sealed Ampule (780200C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malin Edvardsson et al.
Analytical chemistry, 81(1), 349-361 (2008-11-28)
A novel setup was recently developed, combining quartz crystal microbalance with dissipation monitoring (QCM-D) and optical reflectometry for measurements on one and the same surface of, for example, biomolecular adlayers and interactions ( Rev. Sci. Instr. 2008 , 79 075107
J T Elliott et al.
Bioconjugate chemistry, 11(6), 832-841 (2000-11-23)
Two maleimide-containing diacylglycerol derivatives were synthesized to permit the anchoring of short peptides and longer polypeptides to phospholipid bilayers and membranes. The maleimide was introduced at the site normally occupied by a phospholipid headgroup. The first lipid, the dipalmitoyl ester
Xiaoming Zhang et al.
Biochemical and biophysical research communications, 349(3), 920-924 (2006-09-15)
Human serum albumin (HSA) patterns have been successfully fabricated for the deposition of lipid bilayer, 1,2-dimyristoyl-sglycerophosphate (DMPA), by making use of the micro-contact printing (microCP) technique and liposome fusion. Confocal laser scanning microscopy (CLSM) results indicate that lipid bilayer has
K Gradauer et al.
Journal of controlled release : official journal of the Controlled Release Society, 172(3), 872-878 (2013-10-22)
The aim of the present study was the in vivo evaluation of thiomer-coated liposomes for an oral application of peptides. For this purpose, salmon calcitonin was chosen as a model drug and encapsulated within liposomes. Subsequently, the drug loaded liposomes
Bärbel Lorenz et al.
Biophysical chemistry, 150(1-3), 54-63 (2010-03-12)
A versatile model system to study membrane-membrane interactions in great detail is introduced. Based on colloidal probe microscopy with membrane covered spherical probes attached to tip-less cantilevers the interaction forces and adhesion energies are quantified down to single molecule resolution.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service