Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

700367P

Avanti

Tauro-3α,6α,7α,12α-tetrahydroxy bile acid (THBA)

Avanti Research - A Croda Brand

Synonym(s):

Tauro-3α,6α,7α,12α-tetrahydroxy-5ß-cholan-24-oic acid, sodium salt; Tauro-3α,6α,7α,12α-THBA

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$743.00

$743.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$743.00

About This Item

Empirical Formula (Hill Notation):
C26H44NNaO8S
CAS Number:
Molecular Weight:
553.68
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

$743.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700367P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Tauro-3α,6α,7α,12α-tetrahydroxy bile acid (THBA) is a type of bile acid, which is a category of biological molecules produced in the liver from cholesterol. Bile acids play a crucial role in the digestion and absorption of lipids in the small intestine. They are also involved in the regulation of cholesterol levels in the body and can influence liver function and metabolic processes.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service