Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

700078P

Avanti

7α,25-dihydroxycholesterol-d6

Avanti Research - A Croda Brand

Synonym(s):

26,26,26,27,27,27-hexadeutero-7α,25-dihydroxycholesterol ; 111117

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,270.00

$1,270.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$1,270.00

About This Item

Empirical Formula (Hill Notation):
C27H40D6O3
CAS Number:
Molecular Weight:
424.69
UNSPSC Code:
41141804
NACRES:
NA.25

$1,270.00


Please contact Customer Service for Availability

Request a Bulk Order

description

7α,25-dihydroxycholesterol-d6

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700078P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=C[C@H]2O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC(O)(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C27H46O3/c1-17(7-6-12-25(2,3)30)20-8-9-21-24-22(11-14-27(20,21)5)26(4)13-10-19(28)15-18(26)16-23(24)29/h16-17,19-24,28-30H,6-15H2,1-5H3/t17-,19+,20-,21+,22+,23-,24+,26+,27-/m1/s1/i2D3,3D3

InChI key

BQMSKLCEWBSPPY-WWCTWMKLSA-N

General description

7α,25-dihydroxycholesterol (7α,25-OHC) is synthesized by the hydroxylation of cholesterol by the action of enzyme cholesterol 25-hydroxylase (CH25H) and cytochrome P450, family 7, subfamily b, polypeptide 1 (CYP7B1).[1] The catabolic breakdown of 7α,25-OHC to bile acid precursors is catalyzed by the enzymes hydroxy-Δ-5-steroid dehydrogenase, 3β- and steroid Δ-isomerase 7 (HSD3B7).[2] 7α,25-dihydroxycholesterol-d6 is the deuterated form of 7α,25-dihydroxycholesterol.

Application

7α,25-dihydroxycholesterol-d6 has be used as an internal standard in liquid chromatography electrospray ionization tandem mass spectrometric (LC–ESI-MS/MS) analysis of human plasma.[3]

Biochem/physiol Actions

7α,25-dihydroxycholesterol (7α,25-OHC) serves as an endogenous ligand for G-protein-coupled receptor 183 (GPR183) or Epstein-Barr virus-induced gene 2 (EBI2). It plays a key role in the migration of type 3 innate lymphoid cells (ILC3) in the small intestine and colon. 7α,25-OHC mediates immune cell migration functionality by their chemoattractant property.[1] Inhibition of 7α,25-OHC synthesis leads to impairment in the migration of B cells.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (700078P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of structural motifs critical for epstein-barr virus-induced molecule 2 function and homology modeling of the ligand docking site
Zhang L, et al.
Molecular Pharmacology, 82(6), 1094-1103 (2012)
Oxysterol sensing through the receptor GPR183 promotes the lymphoid-tissue-inducing function of innate lymphoid cells and colonic inflammation
Emgaard J, et al.
Immunity, 48(1), 120-132 (2018)
Katharina R Beck et al.
The Journal of steroid biochemistry and molecular biology, 190, 19-28 (2019-03-25)
Oxysterols are cholesterol metabolites derived through either autoxidation or enzymatic processes. They consist of a large family of bioactive lipids that have been associated with the progression of multiple pathologies. In order to unravel (patho-)physiological mechanisms involving oxysterols, it is
Ratna Karuna et al.
Steroids, 99(Pt B), 131-138 (2015-02-17)
We report a straightforward sample preparation procedure and a direct liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method for the analysis of 7alpha,25-dihydroxycholesterol (7α25-OHC) and 7alpha,27-dihydroxycholesterol (7α27-OHC). By applying a slow protein precipitation approach using cold ethanol, we were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service