Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

360130P

Avanti

Huzzah LPA

Avanti Research - A Croda Brand 360130P, powder

Synonym(s):

Huzzah LPA, Huzzah LPA, Human Serum Albumin / Lyso Phosphatidic Acid Complex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (MS)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand 360130P

packaging

1 ea of 1 × (with stopper and crimp cap)

shipped in

dry ice

storage temp.

−20°C

General description

Avanti′s new HSA lipid delivery system, Huzzah improves the cellular delivery of LPA through its conjugation with human serum albumin (HSA), a physiologically relevant carrier protein. This conjugate system eliminates the need for organic solvents, which are well-known to adversely affect viability and the phenotypic characteristics of cells. The Huzzah lipid delivery system is supplied as a lyophilized conjugate which can be directly and easily reconstituted in water or a buffer of choice and is ideal for studying the effects of poorly soluble bioactive molecules.Solubility: Reconstitute lyophilized conjugate in 5 mL of water or other aqueous buffer. This produces a stock solution of 120 - 200 μM LPA*. A short sonication may be necessary to ensure complete dissolution. If necessary, filter sterilize the reconstituted conjugate prior to use in cell culture applications. *Exact concentration will vary from batch to batch. Refer to label for exact product concentration upon reconstitution as determined by mass spectrometric analysis. Molar Ratio: LPA:albumin (2:1) Albumin Biological Characterization: microbial free: endotoxin < 1 IU/mL Albumin Lipid Characterization: ~0.07g/L phospholipids; ~0.02 g/L sphingolipids; ~0.1 g/L fatty acids; and triglycerides and cholesterols below the limit of quantitation of 0.1 g/L and 0.05 g/L

Packaging

10 mL Clear Glass Vial with Stopper and Crimp Cap (360130P-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Huzzah is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service