Skip to Content
MilliporeSigma
All Photos(1)

Documents

330738W

Avanti

Light Bile Acid SPLASH

new

Avanti Research - A Croda Brand

Synonym(s):

Light Bile Acid SPLASH LIPIDOMIX Quantitative Mass Spec Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

biological source

synthetic

description

Technique(s): mass spectrometry

Assay

≥99% (TLC)

packaging

pkg of 1 × 1 ea (330738W-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

This mixture is a complement to Bile Acid SPLASH. It contains the same compounds as Bile Acid SPLASH but without deuterium labels, making this the ideal primary standard mixture for bile acid analysis.

Application

Lipidomics

Packaging

2mL Amber Glass Sealed Ampule (330738W-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service