Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330654

Avanti

GM3 ELISA (Clear)

Avanti Research - A Croda Brand 330654

Synonym(s):

GM3 ELISA, GM3 ELISA Snoopers, Clear, for use in absorbance applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.84

form

liquid

manufacturer/tradename

Avanti Research - A Croda Brand 330654

packaging

1 ea of 1 × (ELISA strip packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

General description

Avanti Lipid Snoopers comprise lipid spotted on solid support and are stable and aid highly sensitive semi-quantitative measurements for lipid detection. Ganglioside GM3 (monosialodihexosylganglioside) are glycosphingolipids (GSLs) which are membrane-bound comprising of a sialic acid residue and an oligosaccharide head structure. It is present in milk and in kidney cells.
ELISA Snoopers are 8-well strips pre-coated with a single high-quality lipid species. They can be used to investigate protein-lipid interactions, and characterize and titrate lipid binding proteins. This format provides the following advantages:
  • quantitative lipid-protein binding studies
  • assemble a 96-well plate using only the lipids of interest for binding studies
  • flexibility - use 8 to 96 wells, and 1 to 12 lipids as needed.

The 8-well strips are available in optically clear, white, or black surfaces for use in colormetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.

Biochem/physiol Actions

Low levels of ganglioside GM3 is implicated in the pathogenesis of rheumatoid arthritis. It modulates immunological functions by inhibiting cytokine production. High GM3 levels are also correlated to the nephrotic syndrome and diabetic nephropathy. Avanti Lipid Snoopers aid in probing protein–lipid interaction by chemiluminescent detection method. Snoopers strips are useful in characterizing lipid-binding antibodies and decoding cell signalling pathways.

Packaging

ELISA Strip Packaged in Foil Pouch (330654-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The missing link-likely pathogenetic role of GM3 and other gangliosides in the development of diabetic nephropathy
Vukovic I, et al.
Kidney & Blood Pressure Research, 40(3), 306-314 (2015)
Ganglioside biochemistry
Kolter, Thomas
ISRN Biochemistry, 2012 (2012)
Ganglioside GM3 has an essential role in the pathogenesis and progression of rheumatoid arthritis
Tsukuda Y, et al.
Testing, 7(6) (2012)
Ganglioside GM3 has an essential role in the pathogenesis and progression of rheumatoid arthritis
Tsukuda Y, et al.
PLoS ONE, 7(6) (2012)
Avanti lipid tools: connecting lipids, technology, and cell biology
Sims KH, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1841(8), 1038-1048 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service