Skip to Content
MilliporeSigma
All Photos(1)

Documents

ATEH998DEB40

AstaTech, Inc.

6-(3-ETHYLUREIDO)PYRIDINE-3-BORONIC ACID PINACOL ESTER

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
291.16
MDL number:

Iupac Name

1-ethyl-3-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)urea

Inchi Code

1S/C14H22BN3O3/c1-6-16-12(19)18-11-8-7-10(9-17-11)15-20-13(2,3)14(4,5)21-15/h7-9H,6H2,1-5H3,(H2,16,17,18,19)

InChI key

ONCZIOGAMMXGRM-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=216868

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D93595


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service