Skip to Content
MilliporeSigma
All Photos(1)

Documents

ATEH95E04DE7

AstaTech, Inc.

2,3-DIFLUORO-5-FORMYLBENZONITRILE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
167.11
MDL number:

Iupac Name

2,3-difluoro-5-formylbenzonitrile

Inchi Code

1S/C8H3F2NO/c9-7-2-5(4-12)1-6(3-11)8(7)10/h1-2,4H

InChI key

KZGAMKRGVDBDBN-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=174866

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C92446


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service