Skip to Content
MilliporeSigma
All Photos(1)

Documents

ATEH93EDCCA7

AstaTech, Inc.

(S)-CYCLOPROPYL(PHENYL)METHANAMINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
147.22
MDL number:

Iupac Name

(S)-cyclopropyl(phenyl)methanamine

Inchi Code

1S/C10H13N/c11-10(9-6-7-9)8-4-2-1-3-5-8/h1-5,9-10H,6-7,11H2/t10-/m1/s1

InChI key

UCRSQPUGEDLYSH-SNVBAGLBSA-N

Purity

95%

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=163745

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=Y13768


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service