Skip to Content
MilliporeSigma
All Photos(1)

Documents

ATE458962856

AstaTech, Inc.

TERT-BUTYL 4-(4-(METHOXYCARBONYL) BENZYL)PIPERAZINE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
334.42
MDL number:

Iupac Name

tert-butyl 4-[4-(methoxycarbonyl)benzyl]-1-piperazinecarboxylate

Inchi Code

1S/C18H26N2O4/c1-18(2,3)24-17(22)20-11-9-19(10-12-20)13-14-5-7-15(8-6-14)16(21)23-4/h5-8H,9-13H2,1-4H3

InChI key

FVMXNXMOUWFRMQ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=71755

Pictograms

Signal Word

Danger

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=20754


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service