Skip to Content
MilliporeSigma
All Photos(1)

Documents

APOH11A9D8BF

Apollo Scientific Ltd

2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
154.17
MDL number:

bp

162-166

Iupac Name

N-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-N-methylamine

Inchi Code

1S/C6H10N4O/c1-4-8-5(7-2)10-6(9-4)11-3/h1-3H3,(H,7,8,9,10)

InChI key

MNDSUSQBIDHEJU-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Product Link

https://store.apolloscientific.co.uk/product/2-methoxy-4-methyl-6-methylamino-135-triazine

Boiling point

162-166

Pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-methoxy-4-methyl-6-methylamino-135-triazine


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service