Skip to Content
MilliporeSigma
All Photos(1)

Documents

APO455836777

Apollo Scientific Ltd

5-Bromo-2-methoxy-3-(trifluoromethyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
256.02
MDL number:

Iupac Name

5-bromo-3-(trifluoromethyl)-2-pyridinyl methyl ether

Inchi Code

1S/C7H5BrF3NO/c1-13-6-5(7(9,10)11)2-4(8)3-12-6/h2-3H,1H3

InChI key

ZIOFHXRZJCIBCO-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/5-bromo-2-methoxy-3-trifluoromethylpyridine

Synonym

5-Bromo-2-methoxy-alpha,alpha,alpha-trifluoro-3-picoline

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-bromo-2-methoxy-3-trifluoromethylpyridine


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service