Skip to Content
MilliporeSigma
All Photos(1)

Documents

APO455835837

Apollo Scientific Ltd

1-Phenyl-5-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
274.63
MDL number:

bp

40-41

Iupac Name

1-phenyl-5-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride

Inchi Code

1S/C11H6ClF3N2O/c12-10(18)8-6-16-17(9(8)11(13,14)15)7-4-2-1-3-5-7/h1-6H

InChI key

FSZPLNSWCMTQRJ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/1-phenyl-5-trifluoromethyl-1h-pyrazole-4-carbonyl-chloride

Flash Point

110/0.2mm

Boiling point

40-41

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/1-phenyl-5-trifluoromethyl-1h-pyrazole-4-carbonyl-chloride


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service