Skip to Content
MilliporeSigma
All Photos(1)

Documents

APO455834566

Apollo Scientific Ltd

2-Fluoro-4-methoxyphenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
247.06
MDL number:

bp

63-66

Iupac Name

2-bromo-1-(2-fluoro-4-methoxyphenyl)ethanone

Inchi Code

1S/C9H8BrFO2/c1-13-6-2-3-7(8(11)4-6)9(12)5-10/h2-4H,5H2,1H3

InChI key

OKLFPKDKQYVRNJ-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/2-fluoro-4-methoxyphenacyl-bromide

Synonym

2-Bromo-2'-fluoro-4'-methoxyacetophenone, 2-Bromo-1-(2-fluoro-4-methoxyphenyl)ethan-1-one, 4-(Bromoacetyl)-3-fluoroanisole

Boiling point

63-66

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-fluoro-4-methoxyphenacyl-bromide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service