Skip to Content
MilliporeSigma
All Photos(1)

Documents

APO455827301

Apollo Scientific Ltd

5-Amino-1-(3,4-dichlorophenyl)-1H-pyrazole-4-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
253.09
MDL number:

bp

170

Iupac Name

5-amino-1-(3,4-dichlorophenyl)-1H-pyrazole-4-carbonitrile

Inchi Code

1S/C10H6Cl2N4/c11-8-2-1-7(3-9(8)12)16-10(14)6(4-13)5-15-16/h1-3,5H,14H2

InChI key

LIBPJAAKKFAEAT-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/5-amino-1-34-dichlorophenyl-1h-pyrazole-4-carbonitrile

Boiling point

170

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-amino-1-34-dichlorophenyl-1h-pyrazole-4-carbonitrile


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service