Skip to Content
MilliporeSigma
All Photos(1)

Documents

AMBH95E088E2

Ambeed, Inc.

tert-Butyldimethyl((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-en-1-yl)oxy)silane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
338.37
MDL number:

Iupac Name

tert-butyl(dimethyl){[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-cyclohexen-1-yl]oxy}silane

Inchi Code

1S/C18H35BO3Si/c1-16(2,3)23(8,9)20-15-12-10-14(11-13-15)19-21-17(4,5)18(6,7)22-19/h10,15H,11-13H2,1-9H3

InChI key

YORHLCKMVFBFLD-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Store in freezer, under -20C

Purity

95%

Country of Origin

CN

Shipping Temperature

Shipped in cold pack

Physical Form

Colorless to yellow liquid or solid

Product Link

https://www.ambeed.com/products/865869-27-8.html

Pictograms

CorrosionExclamation mark

Signal Word

Danger

MSDS

https://www.ambeed.com/products/865869-27-8.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service