Skip to Content
MilliporeSigma
All Photos(1)

Documents

AMBH303C5772

Ambeed, Inc.

6-Methoxy-1,2,3,4-tetrahydroisoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
163.22
MDL number:

Iupac Name

methyl 1,2,3,4-tetrahydro-6-isoquinolinyl ether

Inchi Code

1S/C10H13NO/c1-12-10-3-2-9-7-11-5-4-8(9)6-10/h2-3,6,11H,4-5,7H2,1H3

InChI key

YYTAYINRPUJPNH-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,2-8C

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Form: liquid or solid / Colour: Colorless - Yellow

Product Link

https://www.ambeed.com/products/42923-77-3.html

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service