Key Documents
Z759015
Bollé Sphere safety visor with headgear & brow guard
CE compliant
Synonym(s):
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses
About This Item
UNSPSC Code:
46181802
NACRES:
NB.35
Pricing and availability is not currently available.
feature
CE compliant
packaging
pkg of 1 ea
manufacturer/tradename
Bollé SPHERE
availability
not available in USA
General description
An exceptional field of vision of 180°, the easy to raise headgear and brow guard are adjustable for both height and depth. The Bollé SPHERE adapts itself to any situation, for any use.
  • Ultra enveloping face shield
  • Adjustable headgear and brow guard
  • Polycarbonate visor
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Does the following visor Z759015 Bollé Sphere safety visor with headgear & brow guard protect from the UV light?
    1 answer
    1. This item is not tinted or intended for use as a UV shielding device. The polycarbonate shield offers protection in chemical splash events, cutting or grinding, spraying, or wood and metal work.
      Helpful?