Z757292
Kern ABT analytical balances
model 120-5DM, weighing capacity 42-120 g, AC/DC input 220 - 240 V AC, universal plug set
Synonym(s):
analytical scales
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
stainless steel weighing plate
packaging
pack of 1 ea
manufacturer/tradename
Kern ABT 120-5DNM
measuring range
.01 mg
resolution: 0.1 mg
weighing capacity measuring range
42-120 g
weighing pan W × D
8 cm × 8 cm
Looking for similar products? Visit Product Comparison Guide
General description
The Kern ABT analytical balances are the premium balance model with a high-quality single cell weighing system. It has a unique dosage aid which has a high-stability mode and other filter settings. The simple recipe weighing and documenting is provided with a combined tare/print function, and the simple adding function has auto-tare and automatic subsequent data output after each weighing procedure.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service