Documents
Z756970
Sigma-Aldrich
Aldrich® tubing anchor clips and ties kit
adhesive backs, tubing O.D. 1/8- 3/4 in.
Synonym(s):
hanking ties, laboratory tubing clamps, laboratory tubing clips, tubing clamps, tubing clips, wrap ties
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
tubing O.D.
1/8- 3/4 in.
General description
85 assorted plastic anchor clips with adhesive backs and hanking ties in a plastic storage case for 1/8 in. to 3/4 in. O.D. tubing. Excellent for securing vacuum, gas and liquid lines in fume hoods and around reaction setups, and on benchtops for water lines to filter apparatus, etc.
Kit contents:
  • 30 - Hanking ties, 6 in. L, 10 EA of fine, medium, wide body styles
  • 15 - Top entry adhesive clips, 5 EA, 1/4 , 3/8 ,1/2 in. OD tubing
  • 25 - Side entry adhesive clips, 5 EA, 1/8 , 1/4 , 3/8 , 1/2 , 3/4 in. OD tubing
  • 15 - Adjustable ratcheting clips, 5 EA 1/4, 1/2, 3/4 in. range
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC