Key Documents
Z744039
Sigma-Aldrich
KitAlysis Vials
Synonym(s):
Pack of 96 shell vials (8x30) with stackable tray
Select a Size
1 EACH
$136.00
$136.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EACH
$136.00
About This Item
UNSPSC Code:
12161503
NACRES:
NA.22
$136.00
Please contact Customer Service for Availability
Request a Bulk Order
General description
KitAlysis Vials is a pack of 96 shell vials that are compatible with the KitAlysis High-Throughput Screening Platform. These vials allow the user to screen their own catalysts and ligands using the KitAlysis 24-well Reaction Block.
Choose from one of the most recent versions:
Certificates of Analysis (COA)