Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743807

ART® Non-Filtered Low Retention Tip

HLT 10, size 10 μL, racked, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121603
NACRES:
NB.02
volume:
10 μL
sterility:
sterile
feature:
HLT 10
racked
packaging:
pkg of 5X960 ea
manufacturer/tradename:
Molecular BioProducts 3501-05

sterility

sterile

feature

HLT 10
racked

packaging

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 3501-05

technique(s)

PCR: suitable

size

10 μL

volume

10 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Supplied in environmentally friendly packaging. the non-filtered low retention reload system combines individually packaged blister tubs made of recycled plastic and new hinged racks. The push down blister tub easily inserts the ART barrier tips into the hinged racks providing stackable, space saving racks that offer easy, one-handed use. Empty hinged rack part numbers allow for easy conversion to the ART Reload System. Manufactured according to the industry′s highest quality standards, ART tips are certified as RNase, DNase and Pyrogen free, and are ideal for the pipetting of radioactive samples, nucleic amplification procedures, or any applications where critical sample handling is required. ART™ non-filtered low retention pipette tips ensure reduced sample loss and maximize sample recovery.

Features and Benefits

  • Low retention non-filter tips
  • Tip′s hydrophobic inner surface facilitates accurate data.
  • Hinged racks allow for easy access to the tips.
  • Compatible with radioactive compounds, viscous materials, PCR reagents, and more.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service