Documents
Z742366
2MAG MIXcontrol MTP control device
AC/DC input 100-240 V, UK plug
Synonym(s):
control unit, power bank, power unit
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
material
stainless steel housing
stainless steel plate
feature
4 step power setting (25/50/75/100%)
packaging
pack of 1 ea
manufacturer/tradename
2mag 90250
parameter
0-40 °C temp. limit
100-2000 rpm speed
W × D × H
200 mm × 155 mm × 38 mm
plate size × W × L
× 200 mm × 120 mm, no of position= 66
weight
~1.4 kg
General description
Universal control unit for the connection of one stirring drive MIXdrive MTP, universal 10 watts stirring power, wide speed range from 100 up to 2,000 rpm, jerk-free stirring also at low speeds, 4-step power setting.
Clear digital display for speed- and power settings, SoftStart for reliable centering and safe accelerating of the stirring bar.