Key Documents
Z742307
Horizontal rotisserie for RotoBot Rotating Mixers
holds 6 x 50 mL tubes, pkg of 1 ea
Select a Size
1 EA
$92.80
$92.80
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$92.80
About This Item
UNSPSC Code:
41103801
NACRES:
NB.73
$92.80
Please contact Customer Service for Availability
feature
holds 6 x 50 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific R4040-HZ50
tube diam.
28-30 mm
General description
For higher capacity applications, these holders can be purchased as accessories for the Rotobot and Rotobot Mini rotating mixers. In addition, horizontal holders are available for applications such as Western blotting and hybridization. The unique, sliding attachment allows for quick and easy changing of tube holders.
Legal Information
Rotobot is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)