Documents
Z742253
2MAG distriBOX 4
4-way distributor for the synchronous operation of 2 or 4 MIXdrive stirring drives
Synonym(s):
stirring drive junction box
About This Item
UNSPSC Code:
41123304
NACRES:
NB.72
material
stainless steel
manufacturer/tradename
2mag 49004
cable L
49.0 cm (19.3 in.)
weight
0.48 kg (0.88 lb)
General description
Accessory for use with select 2mag MIXdrive Stirring Drives.
4 fold distributor for synchronized operation of 2 or 4 2mag MIXdrive Stirring Drives. Requires 2 or 4 stirring drives and 1 control unit.
Benefits include identical operating conditions, synchronous speeds, same stirring power, and space saving footprint. Stirring power from control unit will be divided equally to each stirring drive (e.g., 20 watts divided to 10 watts @ each of 2 stirring drives, 40 watts divided to 10 watts @each of 4 stirrings drives, etc.).
Compatible with the following stirring drives: 2mag cuvetteMIXdrive Stirring Drive, 2mag MIXdrive 1 XS (and HT) Stirring Drive, 2mag MIXdrive 1 eco (and HT) Stirring Drive, and 2mag MIXdrive 1 (and HT) Stirring Drive.
Compatible with the following control units: 2mag MIXcontrol 20 (and RS232) Control Unit and 2mag MIXcontrol 40 (and RS232) Control Unit.