Key Documents
Z741837
Extech Portable Sucrose Brix Refractometer (0 to 32%)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.61
feature
(WEEE compliant)
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments RF10
measuring range
0 (sucrose (Brix))
size
6.6 in. × 1.6 in. × 1.2 in., 170 x 40 x 30mm
weight
7 oz200 g
General description
  • Measure the concentration of Sugar, Salt, Lubricants, Antifreeze, Cutting Fluids & more
  • Easy to operate refractometers provide accurate and repeatable measurements on easy to read scales
  • Automatic temperature compensation models available for sugar, salt, lubricants, cutting fluids, and antifreeze
  • Requires only 2 or 3 drops of solution
  • The prism and lens with a simple focus adjustment provides repeatable results
  • Complete with case, calibration screwdriver, and calibration solution
Choose from one of the most recent versions:
Certificates of Analysis (COA)