Key Documents
Z741835
Extech Stem Thermometer, Waterproof
About This Item
UNSPSC Code:
42182209
NACRES:
NB.79
feature
CE compliant
battery included (LR44)
resolution 0.1 °F/C
(WEEE Compliant)
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments 39240
temperature measuring range
-40-392 °F (-40 to 200 °C)
size
5.9 in. × 0.8 in. × 0.7 in., 150 x 20 x 18mm
weight
0.7 oz20 g
Features and Benefits
  • Pocket size with Max/Min recording
  • Measures temperature in frozen solids and liquids
  • 2.75" (70mm) Stainless steel stem
Choose from one of the most recent versions:
Certificates of Analysis (COA)