Z741807
Extech Portable Indoor Air Quality, Carbon Dioxide (CO2) Meter
About This Item
Recommended Products
feature
CE compliant
battery included
WEEE compliant
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments CO240
humidity measuring range
10-90 %RH, accuracy: ±5 %RH, resolution: 0.1 %RH
measuring range
0-9999 ppm, accuracy: ±75 ppm, resolution: 1 ppm (CO2)
temperature measuring range
-10-50 °C, accuracy: ±1.0 °C, resolution: 0.1 °C
size
8.3 in. (211 mm) × 2.4 in. (60 mm) × 1.6 in. (40 mm)
weight
6.4 oz (181.4 g)
battery power
6 AAA
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service