Key Documents
Z741760
Whatman® Polycap HD heavy duty encapsulated filter
Polycap HD 36, pore size 10.0-20.0 μm
Synonym(s):
Whatman filter, Whatman filters, filter Whatman
About This Item
UNSPSC Code:
41104900
Pricing and availability is not currently available.
material
polypropylene housing
polypropylene membrane
sterility
non-sterile
packaging
pkg of 5 ea
manufacturer/tradename
Whatman 2614T
Whatman Article No. 28418923 (US reference)
pore size
10.0-20.0 μm
fitting
inlet 0.125 inch FNPT
outlet 0.125 inch FNPT
General description
Whatman Polycap HD capsule filters are heavy duty polypropylene filters, commonly used in processing applications for general sample preparation; fine filtration; and refiltration for reverse osmosis, ultrafiltration and microfiltration membranes.
Heavy duty polypropylene filter capsule for chemical processing
Whatman Polycap HD capsule filters are used in processing applications to filter buffers, solvents, reagents and emulsions across a wide range of industries including clean air and gas equipment, cosmetics and personal care, food and beverage, inks and pigments, pharmaceuticals, photography, semiconductors and magnetic media.
Chemical processing applications often require a capsule filter to ensure a finishing step that yields a high purity final product. Different solution temperatures and pH values can complicate filtration. Whatman Polycap HD capsule filters are manufactured with polypropylene filter media, support system and housing allow use with a broad range of solutions, pH, and temperatures. Sturdy construction enables high flow rate and high retention capacity for excellent filtrate purity.
Other Notes
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)