Key Documents
Z685216
Replacement HEPA filter for Plas-Labs PCR UV chamber
Select a Size
1 EA
$355.00
$355.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$355.00
About This Item
UNSPSC Code:
41103406
NACRES:
NB.35
$355.00
Please contact Customer Service for Availability
manufacturer/tradename
Plas-Labs 825-HEPA
technique(s)
PCR: suitable
Related Categories
General description
HEPA filter is a replacement for the Plas-Labs PCR UV chamber with HEPA filter, Z566217, Z566225 and Z566233.
Choose from one of the most recent versions:
Certificates of Analysis (COA)