Key Documents
Z685097
Ace sparger with hose barb
L × O.D. 915 mm × 14 mm, porosity 70-100 μm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
manufacturer/tradename
Ace Glass 6452-44
L × O.D.
915 mm × 14 mm
stopcock size
0-5 mm
pore size
70-100 μm porosity
General description
Sparger tube with Ace fritted bottoms are available in various porosities. Top has either 0-3 or 0-5 mm stopcock and a 90 degree side arm hose connection. 10 mm O.D. spargers fit inside #11 Ace-Threds, 14 mm O.D. within #15 Ace- Threds.
Choose from one of the most recent versions:
Certificates of Analysis (COA)