Key Documents
Z682659
Ace drying flask, pear-shaped, with side indents
capacity 500 mL, joint: ST/NS 24/40, glass
Synonym(s):
Aldrich® drying flask, pear-shaped, with side indents
Select a Size
1 EA
$224.00
$224.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$224.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$224.00
Please contact Customer Service for Availability
material
glass
pear-shaped flask
single-neck
Joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Büchi equivalent, 11579
capacity
500 mL
General description
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)