Documents
Z682160
Ace stopcock only, with inlet feed, for condenser "C" and "CR" assemblies
vented, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint ST/NS19/38
manufacturer/tradename
Büchi equivalent, 40627
Related Categories
General description
The Aldrich stopcock fits the Aldrich condenser "C" and "CR" assemblies.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG