Key Documents
Z680575
Pouring rings for Duran® laboratory bottles
PTFE, neck thread GL 80
Synonym(s):
pouring ring
Select a Size
5 EA
$392.00
$392.00
Please contact Customer Service for Availability
Select a Size
Change View
5 EA
$392.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
$392.00
Please contact Customer Service for Availability
material
PTFE
manufacturer/tradename
DURAN®
Schott
General description
The material used for this GLS 80 closure is a special compound based on polyarylsulphone. As a result, the chemical, thermal and mechanical properties of the material are improved compared to polypropylene. Thanks to the shortened triple thread, the DURAN® GLS 80® bottle can be opened or closed with only a three-quarter turn. The seal, which is coated on both sides with PTFE, ensures the bottle can be tightly sealed (cap liner: platinum-cured silicone). A matching PTFE pouring ring is also available, permitting clean, drip-free use.
Features and Benefits
Optional pouring rings prevent drips when pouring from Duran bottles.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)