Documents
Z678090
Finnpipette® F3 variable volume pipette
volume 1-10 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
color-coded pink
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Finn 4640000
volume
1-10 μL
General description
Finnpipette F3 has a double action pipetting button with rotating upper part enabling smooth and steady pipetting action at the same time as preventing accidental volume alterations. Finpipette F3 is equipped with the Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Double action pipetting button with very light and smooth pipetting action
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Color coded for easy tip volume recognition
Compatibility
Uses Finntip 10 or Flex 10
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries