Key Documents
Z673900
Hanna HI 9126 portable pH/ORP meter
About This Item
UNSPSC Code:
41115603
manufacturer/tradename
Hanna HI 9126
mV measuring range
±699 mV, accuracy: 0.2 mV
pH measuring range
-2.00-16.00, accuracy: 0.01
temperature measuring range
-9.9-120 °C, accuracy: ±0.5 °C
General description
  • Exclusive Calibration Check
  • Electrode condition monitoring
  • Large multi-level display LCD
  • On-screen operational guide
  • Waterproof
  • Auto calibration at 1 or 2 points
The HI 9126 is designed to provide laboratory results in a sleek, portable design. Supplied with rechargeable batteries and an inductive docking station for easy recharging. The multi-level LCD displays pH and temperature readings simultaneously, while also displaying the electrode status and operational guides.
Battery Error Prevention System (BEPS) automatically turns the instrument off when battery level is too low to ensure reliable readings.
The HI 9126 can also be used to measure ORP in the mV range with 0.1 mV resolution.
  • Exclusive Calibration Check
  • Electrode condition monitoring
  • Large multi-level display LCD
  • On-screen operational guide
  • Waterproof
  • Auto calibration at 1 or 2 points
Supplied with HI 1230B combination double junction, gel-filled pH electrode, HI 7662 SS temperature probe, electrode holder, pH 4.01 & 7.01 buffer solutions, 100 mL plastic beaker, 4 x 1.2 V AAA rechargeable batteries (up to 2,000 hours life), inductive recharger, instruction manual and rugged carrying case.
Choose from one of the most recent versions:
Certificates of Analysis (COA)