Key Documents
Z671940
KNF LIQUIPORT chemically-resistant diaphragm liquid pumps
AC/DC input 230 V AC, polypropylene (head), flow rate 0.5-3.0 L/min
About This Item
UNSPSC Code:
41105101
material
polypropylene (head)
manufacturer/tradename
KNF NF 300 KT.18 S
parameter
0.5-3.0 L/min flow rate
tube I.D.
12 mm
weight
1.5 kg
output
24 watts (motor)
General description
KNF′s new NF liquid pumps are the convenient way to transfer liquids in the laboratory without the mess of peristaltic tubing. They are stylish and compact, and PTRFE models are highly resistant to chemical attack, prevent contamination f your pumped medium.
The NF100 and NF300 pumps are available in either polypropylene or PTFE head materials, with FFPM valve. You should always check corrosion compatibility charts with the medium you intend to pump.
  • 10 mWg pressure head
  • 3 mWg suction head
  • IP65 protection class
  • Maintenance-free
  • Self priming
  • Dry running
  • Adjustable flow rate
Choose from one of the most recent versions:
Certificates of Analysis (COA)