Key Documents
Z656097
Heidolph Reax 2 overhead shaker
AC/DC input 230 V AC
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
Pricing and availability is not currently available.
feature
CE compliant
manufacturer/tradename
541-21009-00 (EU)
541-21009-03 (UK)
541-21009-04 (CH)
availability
available only in EU
parameter
20-100 rpm speed
General description
  • Superior in hematology, medical diagnostics, materials testing and biochemistry.
  • Comes with Easy-to-change universal adapter for use of various vessels
  • Adapter for use of up to 20 test tubes at the same time (accessory)
  • Comes with non-skid rubber mat
  • Analogue rpm adjustable dial controls
  • Protection class IP 21
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Does this include universal adapter? Also, please provide me full description of the product.
    1 answer
    1. Yes, this product includes a universal adapter. Additional specifications are listed below:
      Helpful?