Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z655279

Hanna TDS standards

TDS 1382 ppm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

manufacturer/tradename

Hanna HI 70032C

TDS measuring range

1382 ppm

Looking for similar products? Visit Product Comparison Guide

General description

Hanna conductivity and TDS standards are supplied in 20 mL sachets to eliminate any chance of cross-contamination and provide the convenience of ready-to-use solutions.

Made using NIST standardized potassium chloride and silver nitrate solutions.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service