Key Documents
Z648477
Quickfit® multi-limb vacuum receiver
Quickfit RA 11/SC, four-limb vacuum receiver
Synonym(s):
RA11/SC
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
female joint 14/23
male joint 19/26
manufacturer/tradename
Quickfit RA 11/SC
availability
not available in ROW (Available only in EU countries)
not available in USA and Canada
General description
When suitably lubricated the receiver adapter may be rotated at the conical joint to divert liquid, in turn, to four attached flasks without disturbing the vacuum in the system. Requires four 100 mL flasks (Z302619, not included) with screw-thread adapter for flexible tubing.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)