Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z645664

Microflex® 93-833 UltraSense nitrile gloves

size XL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181504

material

blue gloves
blue nitrile

size

XL

sterility

non-sterile

feature

CE compliant
disposable
latex free: no
powder-free

packaging

package of 100 ea

manufacturer/tradename

MICROFLEX 93-833-100

L × thickness

9.7 in. (245 mm) × 3.5 mil (0.09 mm)

Looking for similar products? Visit Product Comparison Guide

General description

  • Proprietary, ultra-soft formulation provides a comfortable fit and feel
  • Forms to your hand to allow full range of motion
  • Unique finishing process delivers an outstanding grip for both small utensils and large instruments in wet and dry environments
  • Eliminates the possibility of natural rubber latex allergic reactions

Thickness:
  • Cuff: 0.07 mm (2.7 mil)
  • Palm: 0.09 mm (3.5 mil)
  • Finger: 0.12 mm (4.7 mil)
  • Length: 245 mm (9.7")
A soft nitrile glove with a latex fit! UltraSense has a precise fit, with ultra comfort and unwavering grip, conforming to your hand like latex. Unmatched quality and comfort in an affordable nitrile glove. UltraSense forms to your hand like latex, without the associated risks, and has elasticty unlike any other nitrile.

High quality nitrile UltraSense provides reliable barrier protection against many hazardous and infectious substances.

  • Proprietary, ultra-soft formulation provides a comfortable fit and feel
  • Forms to your hand to allow full range of motion
  • Unique finishing process delivers an outstanding grip for both small utensils and large instruments in wet and dry environments
  • Eliminates the possibility of natural rubber latex allergic reactions

Thickness:
  • Cuff: 0.07 mm (2.7 mil)
  • Palm: 0.09 mm (3.5 mil)
  • Finger: 0.12 mm (4.7 mil)
  • Length: 245 mm (9.7")

Other Notes

Microflex UltraSense is in the process of re-branding. The new product name will become Microflex 93-833. Changes effect name and packaging only. No change in material specifications.

Legal Information

Microflex is a registered trademark of CIT Healthcare LLC
UltraSense is a trademark of Microflex Medical Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service