Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z620440

Schott® narrow mouth reagent bottles

capacity 500 mL, flat head glass stopper, clear glass bottle

Synonym(s):

211654408

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.

material

clear glass bottle
flat head glass stopper
narrow-mouth bottle
round bottle

Joint

joint ST/NS24/29

feature

cap

manufacturer/tradename

DURAN®
Schott 211654408

availability

not available in USA and Canada

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information

DURAN is a registered trademark of DWK Life Sciences
Schott is a registered trademark of Schott AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service