Key Documents
Z602876
Normag reflux boiling splitter
bottom joint: 29/32
About This Item
UNSPSC Code:
12352200
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
bottom joint: 29/32
side joint 14/23
manufacturer/tradename
Normag
availability
not available in USA and Canada
Features and Benefits
With fine adjustment glass needle valve. Installation length 50mm for
thermometers. GL 14 Glass thread cooling water connections. Complete
with 2 GL 14 threaded hose connectors.
Legal Information
Normag is a trademark of Normag Labor- und Prozesstechnik GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)