Z562939
Plas-Labs Nitrogen dry-box glove box
AC/DC input 240 V AC, UK plug, Plas-Labs 850-NB/EXP
Select a Size
$26,460.00
Select a Size
About This Item
$26,460.00
Recommended Products
grade
CE Compliant
manufacturer/tradename
Plas-Labs 850-NB/EXP
H × W × D
26 in. × 41 in. × 26 in., inside dimensions
38 in. × 55 in. × 35 in., outside dimensions
weight
250 lb
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Clear, one piece acrylic top with molded base section
- Double layered, closed cellular neoprene gaskets for airtight seal between top and base
- Adjustable SS clamp compensate for wear
- 8 in. O.D. ports with ambidextrous Hypalon® gloves with SS O-rings
- Transparent transfer chamber (12 L × 11 in. I.D.) with adjustable vacuum gauge
- Gas-drying train with DesiSphere desiccating canisters, self-sealing quick-disconnects enable change of drying train without disturbing internal atmosphere
- Two vacuum/pressure pumps - one for transfer chamber, one for drying train to speed purging
- Fluorescent light system with illuminated controls
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service