Documents
Z557854
Sigma-Aldrich
Sigma-Aldrich® temperature controller for ceramic top hotplate stirrer
About This Item
UNSPSC Code:
41102406
temperature measuring range
accuracy: 1 °C
General description
The digital temperature controller has fuzzy logic control which provides constant feedback to hot plate to ensure perfect temperature control of the medium without overshooting the set temperature. Connects to the stirring hot plates with a DIN 12878 socket. Includes a 250 mm long × 3 mm diameter stainless steel temperature probe.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany