Key Documents
Z541494
Sigma-Aldrich
Aldrich® Spectral Viewer
FT-NMR, standard library, network version
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
General description
The Aldrich Spectral Viewer is an electronic reference book on CD-ROM that contains thousands of spectra from the Aldrich spectral libraries. This easy-to-use software program is more powerful than a printed book, allowing text and data field searching and the manipulation, printing, and exporting of spectra.
Currently, we offer one FT-IR library (over 11,000 compounds), two FT-NMR libraries (over 15,000 compounds), and one ATR-IR library (over 18,000 compounds) to suit your needs. The libraries can be used individually or can be combined to create a powerful, electronic reference. A single user interface handles IR, NMR, and ATR-IR spectra, simplifying the use of the software.
In addition, the Spectral Viewer is expandable so that when new spectral libraries are released, they can be easily added to the software. A student/instructional version is also available with a limited library of FT-NMR, FT-IR, and ATR-IR spectra. The demonstration version is also available with a complete version of the software and a small demonstration database of FT-NMR, FT-IR, and ATR-IR spectra.
For more information on a particular spectral library, please see the individual product listing.
Other Notes
11,800 compounds
System Requirements
600MB hard disk space (server), 30MB hard disk space (client)
Pentium 200MHz, Windows XP / Vista / 7 / 10, CD-ROM drive, 64MB RAM, disk space will vary with libraries installed.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Spectral Viewer is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)