Key Documents
Z531006
Sigma-Aldrich
Aldrich® Liebig condenser
Overall H 300 mm, joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121800
Joint
joint: ST/NS29/32
Overall H
300 mm
General description
Condensers have a standard taper female joint and a lower male drip tip-joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)